Fenantreeni 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

130090050 130090500 130095000
ACRO130090050EA 24.4 EUR
ACRO130090050 ACRO130090500 ACRO130095000
Fenantreeni 97%
Fenantreeni
Kaava: C₁₄H₁₀
Kiehumispiste: 340 °C (1013 hPa)
Sulamispiste: 95…101 °C
Tiheys: 1,063 g/cm³
Leimahduspiste: 153 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001168
CAS-numero: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9,III
Merck Index: 12,07354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92527d856e1fe8582e54bf3812e1580ee0b91b05&vid=d8ec397dac001e49a7958f12aacb577cd11954da&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to grey to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 97 to 101 °C
GC ≥96.0 %
Stabilizer 150 to 400 ppm BHT
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR