3'-Nitroasetofenoni 98%

Toimittaja: Thermo Fisher Scientific

128331000 128335000
ACRO128331000EA 30.3 EUR
ACRO128331000 ACRO128335000
3'-Nitroasetofenoni 98%
3'-Nitroasetofenoni
Kaava: C₈H₇NO₃
Kiehumispiste: 202 °C (1013 hPa)
Sulamispiste: 76…79 °C
Tiheys: 1,433 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007259
CAS-numero: 121-89-1
EINECS: 204-504-3

Order Now

Testitulosten erittely

Appearance Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR