5-Methyl-DL-tryptophan 98%

Toimittaja: Thermo Fisher Scientific

Synonyms: 2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH

127970010 127972500 127970050
ACRO127970010EA 449 EUR
ACRO127970010 ACRO127972500 ACRO127970050
5-Methyl-DL-tryptophan 98%
5-Methyl-DL-tryptophan

CAS No.: 951-55-3

Kaava: C₁₂H₁₄N₂O₂
Sulamispiste: 280…282 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005652
CAS-numero: 951-55-3

Order Now

Testitulosten erittely

Appearance Off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Sulfated ash ≤0.5 %
Appearance of solution (5% soln. in 1 N NaOH) clear, pale yellowish
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR