N-Metyyli-2-pyrrolidoni (NMP) 99%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

127630010 127630025 127630250
ACRO127630010EA 59.1 EUR
ACRO127630010 ACRO127630025 ACRO127630250
N-Metyyli-2-pyrrolidoni (NMP) 99%, extra pure
N-Metyyli-2-pyrrolidoni (NMP)
Kaava: C₅H₉NO
Kiehumispiste: 202 °C (760 mmHg)
Sulamispiste: –24 °C
Tiheys: 1,03 g/cm³
Leimahduspiste: 91 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003193
CAS-numero: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4690 to 1.4710 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥98.5 %
Color scale ≤50 APHA
Specific gravity (20°C) 1.02 to 1.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR