4-Metyylibentsyyliamiini 98%

Toimittaja: Thermo Fisher Scientific
Vaara

126400250 126401000 126400050
ACRO126400250EA 80.8 EUR
ACRO126400250 ACRO126401000 ACRO126400050
4-Metyylibentsyyliamiini 98%
4-Metyylibentsyyliamiini
Kaava: C₈H₁₁N
Kiehumispiste: 195 °C (1013 hPa)
Sulamispiste: 13 °C
Tiheys: 0,95 g/cm³
Leimahduspiste: 75 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008123
CAS-numero: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5360 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR