3-Merkaptopropaanihappo 99+%

Toimittaja: Thermo Fisher Scientific
Vaara

125531000 125535000 125530025
ACRO125531000EA 56.8 EUR
ACRO125531000 ACRO125535000 ACRO125530025
3-Merkaptopropaanihappo 99+%
3-Merkaptopropaanihappo
Kaava: C₃H₆O₂S
Kiehumispiste: 110…111 °C (15 mmHg)
Sulamispiste: 17…19 °C
Tiheys: 1,218 g/cm³
Leimahduspiste: 124 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004897
CAS-numero: 107-96-0
EINECS: 203-537-0
UN: 2922
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
Titration Iodimetric 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR