1-(4-Nitrophenyl)piperazine 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

124940050 124940250 124941000
ACRO124940050EA 31.8 EUR
ACRO124940050 ACRO124940250 ACRO124941000
1-(4-Nitrophenyl)piperazine 98%
1-(4-Nitrophenyl)piperazine
Kaava: C₁₀H₁₃N₃O₂
Sulamispiste: 130…134 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005961
CAS-numero: 6269-89-2
EINECS: 228-443-7
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Yellow to ochre Powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR