Etikkahappo 99.5%, pure

Toimittaja: Thermo Fisher Scientific
Vaara

124040010 124040025 124040250 124040050
ACRO124040010EA 66 EUR
ACRO124040010 ACRO124040025 ACRO124040250 ACRO124040050
Etikkahappo 99.5%, pure
Etikkahappo
Kaava: H₃CCOOH
Kiehumispiste: 118 °C (1013 hPa)
Sulamispiste: 16…16,5 °C
Tiheys: 1,048 g/cm³
Leimahduspiste: 40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3700 to 1.3730 (20°C, 589 nm)
Color scale ≤10 APHA
Titration with NaOH ≥99.4 %
Appearance of solution (EP) Passes test
Identification Passes test (EP, USP)
Related substances ≤0.5 % (total impurities)
Specific optical rotation +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3) (Anhydrous substance)
Acidity (USP) Passes test
Color of solution (USP) Passes test
Chloride (Cl) ≤0.018 %
Sulfate (SO4) ≤0.025 %
Arsenic (As) ≤1 µg/g
Soluble starch, sulfite (USP) Passes test
Dextrin (EP, USP) Passes test
Conductivity ≤20 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR