Bromivety 33% (w/w) in glacial acetic acid, pure

Toimittaja: Thermo Fisher Scientific
Vaara

123180010 123185000 123180025
ACRO123180010EA 165 EUR
ACRO123180010 ACRO123185000 ACRO123180025
Bromivety 33% (w/w) in glacial acetic acid, pure
Bromivety
Kaava: HBr
Tiheys: 1,42 g/cm³
Leimahduspiste: 39 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011323
CAS-numero: 10035-10-6
EINECS: 233-113-0
UN: 2920
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear yellow to yellow-orange Solution (May darken on storage)
Water ≤0.5 % (K.F.)
Titration with NaOH 31.5 to 36.5 % (HBr)
Particle size ≥50 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR