4-Oxopimelic acid 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

122880050 122880250 122881000
ACRO122880050EA 78 EUR
ACRO122880050 ACRO122880250 ACRO122881000
4-Oxopimelic acid 98%
4-Oxopimelic acid
Kaava: C₇H₁₀O₅
Sulamispiste: 142…144 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002798
CAS-numero: 502-50-1
EINECS: 207-941-8

Order Now

Testitulosten erittely

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 141 to 145 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR