1,1,1,3,3,3-Heksametyylidisilatsaani 98%

Toimittaja: Thermo Fisher Scientific
Vaara

120580100 120581000 120585000 120580025
ACRO120580100EA 27.2 EUR
ACRO120580100 ACRO120581000 ACRO120585000 ACRO120580025
1,1,1,3,3,3-Heksametyylidisilatsaani 98%
1,1,1,3,3,3-Heksametyylidisilatsaani
Kaava: (CH₃)₃SiNHSi(CH₃)₃
Kiehumispiste: 125 °C (760 mmHg)
Sulamispiste: –78 °C
Tiheys: 0,76 g/cm³
Leimahduspiste: 20 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008259
CAS-numero: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3,II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4090 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR