Piperidine-N-carbaldehyde 99%

Toimittaja: Thermo Fisher Scientific
Vaara

119730010 119732500
ACRO119730010EA 336 EUR
ACRO119730010 ACRO119732500
Piperidine-N-carbaldehyde 99%
Piperidine-N-carbaldehyde
Kaava: C₆H₁₁NO
Sulamispiste: –31 °C
Tiheys: 1,019 g/cm³
Leimahduspiste: 91 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006483
CAS-numero: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR