Fisetin

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 3,7,3',4'-Tetrahydroxyflavone

119150010 119150050
ACRO119150010EA 169 EUR
ACRO119150010 ACRO119150050
Fisetin
Fisetin
Kaava: C₁₅H₁₀O₆
MW: 286.24 g/mol
Sulamispiste: >330 °C
MDL numero: MFCD00006829
CAS-numero: 528-48-3
EINECS: 208-434-4

Order Now

Testitulosten erittely

Appearance White to amber to dark green Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR