5,5'-ditiobis(2-nitrobentsoehappo) 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

117540050 117540010 117540250
ACRO117540050EA 88 EUR
ACRO117540050 ACRO117540010 ACRO117540250
5,5'-ditiobis(2-nitrobentsoehappo) 99%
5,5'-ditiobis(2-nitrobentsoehappo)
Kaava: C₁₄H₈N₂O₈S₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00007140
CAS-numero: 69-78-3
EINECS: 200-714-4

Order Now

Testitulosten erittely

Appearance Off-white to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 240 to 245 °C
Loss on drying ≤0.5 %
Titration with NaOH ≥98.5 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR