Dimetyylisulfaatti 99+%

Toimittaja: Thermo Fisher Scientific
Vaara

116820010 116821000 116820050
ACRO116820010EA 105 EUR
ACRO116820010 ACRO116821000 ACRO116820050
Dimetyylisulfaatti 99+%
Dimetyylisulfaatti
Kaava: C₂H₆O₄S
Kiehumispiste: 188 °C (760 mmHg)
Sulamispiste: –32 °C
Tiheys: 1,33 g/cm³
Leimahduspiste: 83 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008416
CAS-numero: 77-78-1
EINECS: 201-058-1
UN: 1595
ADR: 6.1,I
Merck Index: 12,03305

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
Acidity ≤0.2 % (As sulfuric acid)
Titration 99.0 to 101.0 % (Hydrolysis)
Barium (Ba) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤200 ppm
Strontium (Sr) ≤1000 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.01 %
Oxidizing substance (as NO3) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR