2,6-Ksylidiini 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

115940050 115941000 115945000 115940025
ACRO115940050EA 35.8 EUR
ACRO115940050 ACRO115941000 ACRO115945000 ACRO115940025
2,6-Ksylidiini 99%
2,6-Ksylidiini
Kaava: C₈H₁₁N
Kiehumispiste: 216 °C (760 mmHg)
Sulamispiste: 10…12 °C
Tiheys: 0,98 g/cm³
Leimahduspiste: 91 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007747
CAS-numero: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Testitulosten erittely

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5591 to 1.5611 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR