4-(Dimetyyliamino)antipyriini 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

115740050 115741000 115740010 115740250
ACRO115740050EA 26.2 EUR
ACRO115740050 ACRO115741000 ACRO115740010 ACRO115740250
4-(Dimetyyliamino)antipyriini 97%
4-(Dimetyyliamino)antipyriini
Kaava: C₁₃H₁₇N₃O
Sulamispiste: 105…110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003142
CAS-numero: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,00495

Order Now

Testitulosten erittely

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Titration with HClO4 ≥96.0 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR