2,3-Dimetoksibentsoehappo 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

115420050 115420250 115421000
ACRO115420050EA 22.7 EUR
ACRO115420050 ACRO115420250 ACRO115421000
2,3-Dimetoksibentsoehappo 99%
2,3-Dimetoksibentsoehappo
Kaava: C₉H₁₀O₄
Kiehumispiste: 314 °C (1013 hPa)
Sulamispiste: 121…124 °C
Tiheys: 1,387 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002432
CAS-numero: 1521-38-6
EINECS: 216-188-4

Order Now

Testitulosten erittely

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 124 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR