2',6'-Dihydroksiasetofenoni 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

114820100 114820500
ACRO114820100EA 70.6 EUR
ACRO114820100 ACRO114820500
2',6'-Dihydroksiasetofenoni 99%
2',6'-Dihydroksiasetofenoni
Kaava: C₈H₈O₃
Kiehumispiste: 296 °C (1013 hPa)
Sulamispiste: 156…159 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002270
CAS-numero: 699-83-2
EINECS: 211-833-6

Order Now

Testitulosten erittely

Appearance Yellow to light beige Powder
Infrared spectrum Conforms
Melting point 155 to 158 °C
HPLC ≥98.5 %
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR