Diethyl ethylmalonate 99%

Toimittaja: Thermo Fisher Scientific

114361000 114365000
ACRO114361000EA 31.2 EUR
ACRO114361000 ACRO114365000
Diethyl ethylmalonate 99%
Diethyl ethylmalonate
Kaava: C₉H₁₆O₄
Kiehumispiste: 75…77 °C (5 mmHg)
Tiheys: 1 g/cm³
Leimahduspiste: 88 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009167
CAS-numero: 133-13-1
EINECS: 205-093-3

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA
Stabilizer 40 to 60 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR