Diethyl (2-bromoethyl)phosphonate 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

114180050 114180250
ACRO114180050EA 66.3 EUR
ACRO114180050 ACRO114180250
Diethyl (2-bromoethyl)phosphonate 97%
Diethyl (2-bromoethyl)phosphonate
Kaava: C₆H₁₄BrO₃P
Kiehumispiste: 75 °C (1 mmHg)
Tiheys: 1,27 g/cm³
Leimahduspiste: >112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000239
CAS-numero: 5324-30-1
EINECS: 226-193-3

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR