2-Chloroethyldiethylammonium chloride 99.5%

Toimittaja: Thermo Fisher Scientific
Vaara

114030010 114030050 114032500 114030025
ACRO114030010EA 124 EUR
ACRO114030010 ACRO114030050 ACRO114032500 ACRO114030025
2-Chloroethyldiethylammonium chloride 99.5%
2-Chloroethyldiethylammonium chloride
Kaava: C₆H₁₅Cl₂N
Sulamispiste: 208…212 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012519
CAS-numero: 869-24-9
EINECS: 212-786-4
UN: 2928
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 212 °C
Loss on drying ≤1 % (105 °C, 3 hrs)
Titration with HClO4 ≥99.4 % (On dry substance)
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm
Sugars, dextrin Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR