Disykloheksyylikarbodi-imidi 99%

Toimittaja: Thermo Fisher Scientific
Vaara

113900010 113901000 113902500 113900050 113900250
ACRO113900010EA 213 EUR
ACRO113900010 ACRO113901000 ACRO113902500 ACRO113900050 ACRO113900250
Disykloheksyylikarbodi-imidi 99%
Disykloheksyylikarbodi-imidi
Kaava: C₁₃H₂₂N₂
Kiehumispiste: 122…124 °C (6 mmHg)
Sulamispiste: 33…35 °C
Tiheys: 0,95 g/cm³ (40 °C)
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011659
CAS-numero: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1,II
Merck Index: 13,03123

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline mass or waxy solid
Infrared spectrum Conforms
Melting point 33 to 35 °C
GC ≥98.5 %
Appearance of solution (10% in acetone) clear colorless to yellowish
Color scale ≤25 APHA (ASTM D 1209)
Bromine number ≤5 g Br/100 g (ASTM D 1159)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR