Adiponitrile 99%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Hexanedinitrile

113830010 113831000
ACRO113830010EA 99.8 EUR
ACRO113830010 ACRO113831000
Adiponitrile 99%
Adiponitrile
Kaava: C₆H₈N₂
Kiehumispiste: 296 °C (1013 hPa)
Sulamispiste: 1…3 °C
Tiheys: 0,95 g/cm³
Leimahduspiste: 163 °C
MDL numero: MFCD00001975
CAS-numero: 111-69-3
EINECS: 203-896-3
UN: 2205
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
GC ≥98.5 %
Microbial contamination (Salmonella) Absent in 25 g
pH 6 to 8 (50 % suspension in water)
Sulfite (as SO2) ≤10 ppm
Arsenic (As) ≤0.07 ppm
Cadmium (Cd) ≤0.05 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤0.1 ppm
Mercury (Hg) ≤0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR