3,5-Dikloorianiliini 98%

Toimittaja: Thermo Fisher Scientific
Vaara

113121000 113125000
ACRO113121000EA 58.1 EUR
ACRO113121000 ACRO113125000
3,5-Dikloorianiliini 98%
3,5-Dikloorianiliini
Kaava: C₆H₅Cl₂N
Kiehumispiste: 260 °C (741 mmHg)
Sulamispiste: 49…53 °C
Tiheys: 1,58 g/cm³
Leimahduspiste: 133 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007774
CAS-numero: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance White to brown Crystals (May darken on storage)
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥97.5 %
Fluorescence max 565 nm (in Methanol)
Optimum lasing 588 nm (in Methanol)
Molar absorption coefficient 106000 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR