3,5-Di-tert-butylpyrocatechol 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

112970250 112970050
ACRO112970250EA 91.7 EUR
ACRO112970250 ACRO112970050
3,5-Di-tert-butylpyrocatechol 99%
3,5-Di-tert-butylpyrocatechol
Kaava: C₁₄H₂₂O₂
Kiehumispiste: 310 °C (1013 hPa)
Sulamispiste: 97…100 °C
MDL numero: MFCD00008819
CAS-numero: 1020-31-1
EINECS: 213-816-9

Order Now

Testitulosten erittely

Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 97 to 100 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR