Dibenzyl phosphonate 90+%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Varoitus

112660250 112660050 112661000
ACRO112660250EA 88.4 EUR
ACRO112660250 ACRO112660050 ACRO112661000
Dibenzyl phosphonate 90+%, Technical Grade
Dibenzyl phosphonate
Kaava: C₁₄H₁₅O₃P
Kiehumispiste: 110…120 °C (,1 mmHg)
Tiheys: 1,18 g/cm³
Leimahduspiste: >110 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00004774
CAS-numero: 17176-77-1
EINECS: 241-226-1

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5550 (20°C, 589 nm)
NMR ≥90.0 %
UV at 280 nm A: ≤0.05
pH 5.5 to 8.0 (5 % in water)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR