Cystamine dihydrochloride 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

111770250 111771000 111775000
ACRO111770250EA 49.5 EUR
ACRO111770250 ACRO111771000 ACRO111775000
Cystamine dihydrochloride 97%
Cystamine dihydrochloride
Kaava: C₄H₁₂N₂S₂·2HCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012905
CAS-numero: 56-17-7
EINECS: 200-260-7

Order Now

Testitulosten erittely

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Additional info oxalic acid: < 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR