Cyclohexylacetic acid 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

111270050 111270250 111271000
ACRO111270050EA 41.6 EUR
ACRO111270050 ACRO111270250 ACRO111271000
Cyclohexylacetic acid 98+%
Cyclohexylacetic acid
Kaava: C₈H₁₄O₂
Kiehumispiste: 242…244 °C (760 mmHg)
Sulamispiste: 28…32 °C
Tiheys: 1,007 g/cm³
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001518
CAS-numero: 5292-21-7
EINECS: 226-132-0

Order Now

Testitulosten erittely

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 27 to 31 °C
Titration with NaOH ≥98.0 %
Identification Passes test
Sensitivity (to amino acids) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR