m-Kresoli 99%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene

110580010 110585000 110580025 110580100 110581000
ACRO110580010EA 197 EUR
ACRO110580010 ACRO110585000 ACRO110580025 ACRO110580100 ACRO110581000
m-Kresoli 99%
m-Kresoli
Kaava: CH₃C₆H₄OH
Kiehumispiste: 203 °C (760 mmHg)
Sulamispiste: 8…10 °C
Tiheys: 1,03 g/cm³
Leimahduspiste: 86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002302
CAS-numero: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1,II
Merck Index: 13,02604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless to light red Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR