2-Bromethylamine hydrobromide 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

106930250 106931000 106930010 106930025 106932500 106935000
ACRO106930250EA 31.1 EUR
ACRO106930250 ACRO106931000 ACRO106930010 ACRO106930025 ACRO106932500 ACRO106935000
2-Bromethylamine hydrobromide 99%
2-Bromethylamine hydrobromide
Kaava: BrCH₂CH₂NH₃Br
Sulamispiste: 172…176 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012886
CAS-numero: 2576-47-8
EINECS: 219-924-2

Order Now

Testitulosten erittely

Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR