3-Bromibentsonitriili 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: m-Bromobenzonitrile

106700100 106700025 106701000
ACRO106700100EA 66.3 EUR
ACRO106700100 ACRO106700025 ACRO106701000
3-Bromibentsonitriili 99%
3-Bromibentsonitriili
Kaava: C₇H₄BrN
MW: 182.02 g/mol
Kiehumispiste: 225 °C (760 mmHg)
Sulamispiste: 37…41 °C
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001796
CAS-numero: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1 III

Order Now

Testitulosten erittely

Appearance White to light yellow to beige Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR