3'-Bromiasetofenoni 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

106580250 106580050 106581000
ACRO106580250EA 38.5 EUR
ACRO106580250 ACRO106580050 ACRO106581000
3'-Bromiasetofenoni 97%
3'-Bromiasetofenoni
Kaava: C₈H₇BrO
Kiehumispiste: 79…81 °C (2 mmHg)
Tiheys: 1,49 g/cm³
Leimahduspiste: >112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000083
CAS-numero: 2142-63-4
EINECS: 218-396-0

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5749 to 1.5769 (20°C, 589 nm)
GC ≥96.0 %
Residue after ignition ≤0.3 %
Specific optical rotation -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR