4-Anisoyylikloridi 99%

Toimittaja: Thermo Fisher Scientific
Vaara

104850050 104851000 104855000
ACRO104850050EA 29 EUR
ACRO104850050 ACRO104851000 ACRO104855000
4-Anisoyylikloridi 99%
4-Anisoyylikloridi
Kaava: C₈H₇ClO₂
Kiehumispiste: 145 °C (14 mmHg)
Sulamispiste: 22 °C
Tiheys: 1,26 g/cm³
Leimahduspiste: 87 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000687
CAS-numero: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8,II
Merck Index: 13,00676

Order Now

Testitulosten erittely

Appearance Clear colorless to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5820 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥96.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR