m-Anisidiini 99%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 3-Anisidiini, 3-Metoksianiliini

104820050 104821000 104822500 104820010
ACRO104820050EA 35 EUR
ACRO104820050 ACRO104821000 ACRO104822500 ACRO104820010
m-Anisidiini 99%
m-Anisidiini
Kaava: C₇H₉NO
Kiehumispiste: 250…251 °C (760 mmHg)
Sulamispiste: -2 °C
Tiheys: 1,102 g/cm³
Leimahduspiste: 126 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007783
CAS-numero: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Testitulosten erittely

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5820 (20°C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR