3-Aminopyridiini 99%

Toimittaja: Thermo Fisher Scientific
Vaara

104560250 104561000 104565000
ACRO104560250EA 35.1 EUR
ACRO104560250 ACRO104561000 ACRO104565000
3-Aminopyridiini 99%
3-Aminopyridiini
Kaava: C₅H₆N₂
Kiehumispiste: ∼248 °C (1013 hPa)
Sulamispiste: 60…63 °C
Leimahduspiste: 124 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006400
CAS-numero: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472

Order Now

Testitulosten erittely

Appearance Off-white to brown Crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 60 to 63 °C
GC ≥98.5 %
Moisture ≤3.0 % (as packaged)
Viscosity 0.2 to 0.6 s
Free acid (CH3COOH) ≤0.06 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR