(±)-3-Amino-1,2-propaanidioli 98%

Toimittaja: Thermo Fisher Scientific
Vaara

104420250 104421000 104420050 104422500
ACRO104420250EA 83.6 EUR
ACRO104420250 ACRO104421000 ACRO104420050 ACRO104422500
(±)-3-Amino-1,2-propaanidioli 98%
(±)-3-Amino-1,2-propaanidioli
Kaava: C₃H₉NO₂
Kiehumispiste: 264…265 °C (739 mmHg)
Tiheys: 1,17 g/cm³
Leimahduspiste: 155 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008140
CAS-numero: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4940 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR