4-Aminomorpholine 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: N-Aminomorpholine

104110250 104110050
ACRO104110250EA 221 EUR
ACRO104110250 ACRO104110050
4-Aminomorpholine 95%
4-Aminomorpholine
Kaava: C₄H₁₀N₂O
Tiheys: 1,05 g/cm³
Leimahduspiste: 58 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006174
CAS-numero: 4319-49-7
EINECS: 224-347-4
UN: 1993
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR