4-Klooriantraniilihappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 2-Amino-4-chlorobenzoic acid

103320250 103321000
ACRO103320250EA 55.8 EUR
ACRO103320250 ACRO103321000
4-Klooriantraniilihappo 98%
4-Klooriantraniilihappo
Kaava: C₇H₆ClNO₂
Kiehumispiste: 330 °C (1013 hPa)
Sulamispiste: 231…235 °C
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007778
CAS-numero: 89-77-0
EINECS: 201-938-5

Order Now

Testitulosten erittely

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 231 to 235 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR