N-Allylaniline 95%

Toimittaja: Thermo Fisher Scientific
Vaara

102870250 102871000 102870050
ACRO102870250EA 163 EUR
ACRO102870250 ACRO102871000 ACRO102870050
N-Allylaniline 95%
N-Allylaniline
Kaava: C₉H₁₁N
Kiehumispiste: 218…220 °C (760 mmHg)
Tiheys: 0,982 g/cm³
Leimahduspiste: 89 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008638
CAS-numero: 589-09-3
EINECS: 209-633-9
UN: 2810
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5620 to 1.5660 (20°C, 589 nm)
Titration with HClO4 ≥94 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.001 %
Iron (Fe) ≤0.002 %
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.05 % (in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR