Methyl hydrogen adipate 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

102820050 102820250 102821000
ACRO102820050EA 20.4 EUR
ACRO102820050 ACRO102820250 ACRO102821000
Methyl hydrogen adipate 97%
Methyl hydrogen adipate
Kaava: C₇H₁₂O₄
Kiehumispiste: 162 °C (10 mmHg)
Sulamispiste: 8…9 °C
Tiheys: 1,08 g/cm³
Leimahduspiste: >109 °C
MDL numero: MFCD00004418
CAS-numero: 627-91-8
EINECS: 211-019-0

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR