Avantor® ACE®, HPLC Precolumn Filters
Toimittaja: Avantor
ACE-HP210EA 384 EUR
ACE-HP210 ACE-HP205 ACE-CS310 ACE-CS305 ACE-CS210 ACE-CS205
Avantor® ACE®, HPLC Precolumn Filters
Suodattimet Esikolonnit ja fritit
These precolumn filters with PEEK body and stainless steel frit provide protection for analytical and microbore columns. They connect directly to the column without the need for an additional holder.
  • Universal filters, suitable for all HPLC columns
  • Pressure rated to 350 bar (5000 psi)
  • Reusable
Analytical filters are available with 0,5 and 2 µm stainless steel frit options (recommended for 3 and 5 µm columns respectively). Microbore filters have a 0,5 µm stainless steel frit and offer lower dispersion than analytical filters. They are recommended forMore Product Information