Avantor® ACE®, HPLC Column Connectors, Finger-Tight
Toimittaja: Avantor
ACE-CC10EA 114 EUR
ACE-CC10 ACE-CC
Avantor® ACE®, HPLC Column Connectors, Finger-Tight
Kiinnitykset ja liittimet
PEEK HPLC column inlet / outlet connectors. These column connectors are designed for use at both the column inlet and outlet ends. They are suitable for use with ¹/₁₆" ext.Ø tubing and all manufacturers' HPLC systems and all brands of HPLC columns.
  • Universal fitting for HPLC column connections
  • Pressure rated to 350 bar (5000 psi)
  • Reusable
  • Inert and biocompatible, excellent chemical resistance