Avantor® ACE® Phenyl-300, HPLC Columns, 5 µm
Toimittaja: Avantor
ACE-225-0246EA 766 EUR
ACE-225-0246 ACE-225-0502 ACE-225-0503 ACE-225-0510 ACE-225-0520 ACE-225-0546 ACE-225-1002 ACE-225-1003 ACE-225-1004 ACE-225-1010 ACE-225-1020 ACE-225-1046 ACE-225-1220 ACE-225-1502 ACE-225-1504 ACE-225-1510 ACE-225-1520 ACE-225-1546 ACE-225-2501 ACE-225-2502 ACE-225-2510 ACE-225-2520 ACE-225-2546 ACE-225-7520 ACE-225-0103GD ACE-225-0110GD ACE-225-7530
Avantor® ACE® Phenyl-300, HPLC Columns, 5 µm
Kromatografiakolonnit
These ultra inert Avantor® ACE® Phenyl-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes available (3 µm, 5 µm and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Excellent peak shape even at extremely low levels of additives