Avantor® ACE® Phenyl, HPLC Columns, 10 µm
Toimittaja: Avantor
ACE-135-0510EA 0 EUR
ACE-135-0510 ACE-135-0520 ACE-135-0530 ACE-135-1010 ACE-135-1020 ACE-135-1030 ACE-135-1220 ACE-135-1510 ACE-135-1520 ACE-135-1530 ACE-135-2510 ACE-135-2520 ACE-135-2530 ACE-135-2546 ACE-135-7520 ACE-135-0103GD ACE-135-7530 ACE-135-3020 ACE-135-3030 ACE-135-3039 ACE-135-3004
Avantor® ACE® Phenyl, HPLC Columns, 10 µm
Kromatografiakolonnit
These ultra inert Avantor® ACE® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Offers strong π-π interactions
  • Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
  • Wide range of particle sizes available (2 µm, 3 µm, 5 µm and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the