Avantor® ACE® C18-AR, HPLC Columns, 5 µm
Toimittaja: Avantor
ACE-129-3502EA 2470 EUR
ACE-129-3502 ACE-129-7520 ACE-129-7530 ACE-129-7546 ACE-129-0102GD ACE-129-0103GD ACE-129-0110GD ACE-129-0302 ACE-129-0502 ACE-129-0503 ACE-129-0510 ACE-129-0520 ACE-129-0530 ACE-129-0546 ACE-129-1002 ACE-129-1003 ACE-129-1004 ACE-129-1008 ACE-129-1010 ACE-129-1020 ACE-129-1030 ACE-129-1046 ACE-129-1220 ACE-129-1246 ACE-129-1502 ACE-129-1503 ACE-129-1504 ACE-129-1510 ACE-129-1520 ACE-129-1530 ACE-129-1546 ACE-129-2502 ACE-129-2503 ACE-129-2504 ACE-129-2508 ACE-129-2510 ACE-129-2520 ACE-129-2530 ACE-129-2546 ACE-129-3020 ACE-129-3030 ACE-129-3004 ACE-129-25005S ACE-129-0120GD ACE1292546MVK
Avantor® ACE® C18-AR, HPLC Columns, 5 µm
Kromatografiakolonnit
The Avantor® ACE® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and