Anti-SYN1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB9702
ABNOPAB9702EA 523 EUR
ABNOPAB9702
Anti-SYN1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against native SYN1.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:2000; Immunofluorescence: 1:2000; Immunoprecipitation: 1 ?g/200 ?g lysate ; Western Blot: 1:1000

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR