Anti-GJA1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB9652
ABNOPAB9652EA 585 EUR
ABNOPAB9652
Anti-GJA1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gja1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR