Anti-LOC677879 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB8949
ABNOPAB8949EA 523 EUR
ABNOPAB8949
Anti-LOC677879 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of Fel D1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500

Type: Primary
Antigen: LOC677879
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR