Anti-DAZL Goat Polyclonal Antibody

Toimittaja: Abnova
PAB6834
ABNOPAB6834EA 523 EUR
ABNOPAB6834
Anti-DAZL Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody raised against synthetic peptide of DAZL.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-0.3 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-3 ?g/ml

Type: Primary
Antigen: DAZL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR